missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (APC)

Catalog No. 501121549
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121549 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121549 Supplier Invitrogen™ Supplier No. SA1005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, (APC)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

Specifications

Concentration 0.1 mg/mL or 0.2 mg/mL
Conjugate APC
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 0.5 mL
Content And Storage Contains 1 vial of streptavidin, APC conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 650/660
Flow Cytometer Laser Lines 633 or 635
Shipping Condition Wet Ice
Form Solution
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye APC (Allophycocyanin)
Show More Show Less
What is the molecular weight of Streptavidin, (APC) (Cat. No. SA1005)?

The Streptavidin, (APC) (Cat. No. SA1005) conjugate has not been purified to remove multimers of streptavidin and APC. The molecular weights of streptavidin and APC are approximately 53 and 104 kDa, respectively. Thus, the conjugate will have a mixture of molecular weights starting at approximately 157 kDa for the 1:1 conjugate plus 210 kDa, 261 kDa, etc. for the multimers.

For Research Use Only. Not for use in diagnostic procedures.