missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin - AP

Catalog No. 434322
Encompass
Change view
Click to view available options
Quantity:
3 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
434322 3 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 434322 Supplier Invitrogen™ Supplier No. 434322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin - AP

Reactivity: AP-SA reacts with biotin.

Applications: Immunohistochemistry, Western blotting, and ELISA

Specifications

Concentration 1.5 mg/mL
Conjugate AP
Quantity 3 mg
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid
Product Type Streptavidin Conjugate (fluorescent)
Label or Dye AP (Alkaline Phosphatase)

For Research Use Only. Not for use in diagnostic procedures.