missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Alexa Fluor™ 750—Allophycocyanin Conjugate (Alexa Fluor™ 750—Allophycocyanin Streptavidin)

Catalog No. 501121178
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121178 Supplier Invitrogen™ Supplier No. S21008

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, Alexa Fluor™ 750—Allophycocyanin Conjugate (Alexa Fluor™ 750—Allophycocyanin Streptavidin)

The streptavidin, Alexa Fluor™ 750-allophycocyanin (APC) streptavidin tandem conjugate has excitation and emission maxima at ∼650/779 and is widely used to detect biotinylated probes by flow cytometry in multicolor applications following excitation at 633 nm or 647 nm.

Related Links:

Specifications

Concentration 1 mg/mL
Conjugate APC-Alexa Fluor 750
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 100 μL
Content And Storage Contains 1 vial of streptavidin APC-Alexa Fluor™ 750 conjugate (1 mg/mL).

Store in refrigerator (2–8°C) and protect from light.
Excitation/Emission 633 or 635/775
Flow Cytometer Laser Lines 633 or 635
Shipping Condition Wet Ice
Form Solution
Product Line Alexa Fluor
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye APC-Alexa Fluor 750
Show More Show Less
Can I use my tandem dye antibody (e.g., Alexa Fluor 610-R-phycoerythrin) for microscopy?

Tandem dyes are not recommended for imaging applications. Both R-PE or APC tend to photobleach quickly and cannot be archived. Any unfolding of the phycobiliprotein would result in loss of fluorescence. They work great in flow cytometry applications.

How should I store my allophycocyanin (APC)-conjugated antibody?

Allophycocyanin-conjugated antibodies must be stored at 2-8 degrees C. Antibodies conjugated to phycobiliproteins (APC, R-PE) must never be frozen. Once frozen, even briefly, they are no longer usable.

For Research Use Only. Not for use in diagnostic procedures.