missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Alexa Fluor™ 647—R-Phycoerythrin Conjugate (Alexa Fluor™ 647—R-Phycoerythrin Streptavidin)

Catalog No. 501137612
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501137612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501137612 Supplier Invitrogen™ Supplier No. S20992

Streptavidin, Alexa Fluor™ 647—R-Phycoerythrin Conjugate (Alexa Fluor™ 647—R-Phycoerythrin Streptavidin)

The streptavidin, Alexa Fluor™ 647-R-phycoerythrin streptavidin tandem conjugate has excitation and emission maxima at ∼496,565/667 and is widely used to detect biotinylated probes by flow cytometry in multicolor applications using a single excitation wavelength, 488 nm. The Alexa Fluor™ 647 conjugates of R-PE have emission spectra almost identical to those of Cy5-R-PE conjugates but tend to have more intesne long-wavelength emission and to require less compensation in the R-PE channel.

View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.

Specifications

Concentration 1 mg/mL
Conjugate PE-Alexa Fluor 647
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 100 μL
Content And Storage Contains 1 vial of streptavidin R-PE-Alexa Fluor™ 647 conjugate (1 mg/mL).

Store in refrigerator (2–8°C) and protect from light.
Excitation/Emission 488 or 561/668
Flow Cytometer Laser Lines 633/635, 488 or 561
Shipping Condition Wet Ice
Form Solution
Product Line Alexa Fluor
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye PE-Alexa Fluor 647
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.