Learn More
Invitrogen™ Streptavidin, Alexa Fluor™ 610—R-Phycoerythrin Conjugate (Alexa Fluor™ 610—R-Phycoerythrin Streptavidin)
Description
The streptavidin, Alexa Fluor™ 610-R-phycoerythrin streptavidin tandem conjugate has excitation and emission maxima at ∼496,565/627 and is widely used to detect biotinylated probes by flow cytometry. Not only are the Alexa Fluor™ 610 conjugates of R-PE are more fluorescent than the commercially available Texas Red™ conjugates of R-PE, the fluorescent emission of Alexa Fluor™ 610-R-PE tandem conjugates is shifted to somewhat longer wavelengths than is the emission of Texas Red™-R-PE conjjgates, resulting in better separation from the emission of R-PE.
Related Links:
Specifications
Specifications
| Concentration | 1 mg/mL |
| Conjugate | PE-Alexa Fluor 610 |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis |
| Quantity | 100 μL |
| Content And Storage | Contains 1 vial of streptavidin R-PE-Alexa Fluor™ 610 conjugate (1 mg/mL). Store in refrigerator (2–8°C) and protect from light. |
| Excitation/Emission | 488 or 561/630 |
| Flow Cytometer Laser Lines | 488 or 561 |
| Shipping Condition | Wet Ice |
| Form | Solution |
| Show More |
Frequently Asked Questions (FAQs)
Tandem dyes are not recommended for imaging applications. Both R-PE or APC tend to photobleach quickly and cannot be archived. Any unfolding of the phycobiliprotein would result in loss of fluorescence. They work great in flow cytometry applications.
For Research Use Only. Not for use in diagnostic procedures.