missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Alexa Fluor™ 532 conjugate

Catalog No. S11224A
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11224A 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11224A Supplier Invitrogen™ Supplier No. S11224

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, Alexa Fluor™ 532 conjugate

Alexa Fluor™ 532 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor™ 532 Streptavidin Conjugates:
  • Alexa Fluor™ 532 streptavidin conjugate has Ex/Em maxima of ∼ (530/554)
  • Bright, photostable fluorescence
  • High solubility in aqueous solutions
  • Available in multiple colors
  • Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

    Properties of Alexa Fluor™ Dyes
    Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

    Blocking Endogenous Biotin
    Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • Specifications

    Conjugate Alexa Fluor 532
    Quantity 1 mg
    Content And Storage Store in freezer (-5 to -30°C) and protect from light.
    Shipping Condition Room Temperature
    Form Solid
    Product Line Alexa Fluor
    Product Type Streptavidin Conjugate (fluorescent)
    Label or Dye Alexa Fluor Dyes
    I am planning to use a fluorescent streptavidin labeled conjugate. What are the storage conditions and shelf life for the lyophilized powder and reconstituted solution?

    In the lyophilized powder form, the fluorescent streptavidin labeled conjugate is stable for six months when stored at -20 degrees C, desiccated, and protected from light. The reconstituted solution is stable for approximately six months when stored at 4 degrees C, protected from light, with the addition of sodium azide to a final concentration of 5 mM or thimerosal to 0.2 mM. For longer storage, we recommend dividing the solution into aliquots and freezing at -20 degrees C, protected from light. Avoid repeated freezing and thawing of the solution.

    I am planning to use a fluorescent streptavidin labeled conjugate. How should I prepare the working solution of the conjugate?

    The fluorescent streptavidin labeled conjugate solution can be made by dissolving the powder in 0.5-1.0 mL of PBS or other suitable buffer. For details, please refer to page 4 of the "Streptavidin and Fluorescent Conjugates of Streptavidin" manual (https://assets.fishersci.com/TFS-Assets/LSG/manuals/mp00888.pdf).

    For Research Use Only. Not for use in diagnostic procedures.