missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Alexa Fluor™ 405 conjugate

Catalog No. S32351
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S32351 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S32351 Supplier Invitrogen™ Supplier No. S32351

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, Alexa Fluor™ 405 conjugate

Our streptavidin, Alexa Fluor 405 conjugate is a bright, photostable probe with excitation/emission maxima of 402 and 421 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.

View a selection guide for all Alexa Fluor™ streptavidin and avidin conjugates.

Specifications

Conjugate Alexa Fluor 405
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 1 mg
Content And Storage Contains 1 vial of streptavidin Alexa Fluor™ 405 conjugate.

Store in freezer (-5 to -30°C) and protect from light.
Excitation/Emission 402/421
Flow Cytometer Laser Lines 405
Shipping Condition Room Temperature
Form Solid
Product Line Alexa Fluor
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye Alexa Fluor 405
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.