missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, Alexa Fluor™ 350 conjugate

Catalog No. s11249
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11249 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. S11249 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' S11249

Alexa Fluor™ 350 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye).

Alexa Fluor™ 350 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.

Important Features of Alexa Fluor™ 350 Streptavidin Conjugates:
Alexa Fluor™ 350 streptavidin conjugate has Ex/Em maxima of ∼ (346/442)
Bright, photostable fluorescence
High solubility in aqueous solutions
Available in multiple colors
Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

Properties of Alexa Fluor™ Dyes
Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.

Consult user Manual for solubility instructions.

Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit (Cat. No. E21390) to minimize this interference.

Specifications

Conjugate Alexa Fluor 350
Quantity 1 mg
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Shipping Condition Room Temperature
Form Solid
Product Line Alexa Fluor
Product Type Streptavidin Conjugate (fluorescent)
Label or Dye Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.