missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, solid

Catalog No. S888
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S888 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S888 Supplier Invitrogen™ Supplier No. S888

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.

  • A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
  • Binding activity of streptavidin is determined to be >11 units per mg protein
  • One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)
  • Store at 4°C

Specifications

Conjugate Unconjugated
Quantity 5 mg
Content And Storage Storage Upon Receipt:≤-20°C, desiccate
Shipping Condition Room Temperature
Form Solid
Product Type Streptavidin Conjugate (fluorescent)
How many biotin binding-sites are there per streptavidin molecule?

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

Is Streptavidin (Cat. No. S888, 434302, and 434301) RNAse-free?

Although we utilize manufacturing methods that minimize the introduction of any contaminants such as RNases, we do not process this product to specifically remove or inactivate RNases and do not test for RNase activity.

For Research Use Only. Not for use in diagnostic procedures.