missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, lyophilized powder DFS Item

A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.

$219.00 - $752.00

Specifications

Conjugate Unconjugated
Content And Storage Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C.
Shipping Condition Wet Ice
Form Lyophilized
Product Type Streptavidin Conjugate (fluorescent)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
434302 DFS Item
View Documents
Invitrogen™
434302
5 mg
Each for $752.00
Only null left
 
434301 DFS Item
View Documents
Invitrogen™
434301
1 mg
Each for $219.00
Only null left
 
Description

Description

  • A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
  • Binding activity of streptavidin is determined to be >11 units per mg protein
  • One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)
  • Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4
  • Reconstitute with 1 mL of distilled or deionized water
  • Store at 4°C
Specifications

Specifications

Unconjugated
Wet Ice
Streptavidin Conjugate (fluorescent)
Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C.
Lyophilized
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.