missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin

Catalog No. 09920068
Change view
Click to view available options
Packaging Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
09920068 10 mL
09920069 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09920068 Supplier Cytiva Supplier No. 29401324

Chromatography resin for purification of recombinant proteins tagged with a Strep-tag™ II and Twin-Strep-tag™.

  • Highly pure Strep-tag II and Twin-Strep-tag recombinant proteins
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration
  • Compatible with a wide range of detergents, denaturants, and other additives

Specifications

For Use With (Application) Purification of recombinant proteins tagged with Strep-tag II and Twin-Strep-tag
Stability Stable in all commonly used aqueous buffers
Storage Requirements 2 to 8°C in 20% ethanol
Binding Capacity Approx. 10 mg protein with Strep-tag II or Twin-Strep-tag/mL of resin
Particle Size 34 μm
Matrix Rigid, cross-linked agarose, spherical
pH Range 6 to 10 pH
Density (Metric) Approx. 5 mg/mL resin
Ligand Type Strep-Tactin XT
Packaging Quantity 10 mL