missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STRBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554256 Supplier Invitrogen™ Supplier No. PA554256

Rabbit Polyclonal Antibody

Immunogen sequence: CLASGILLPG GPGLHDPCER DPTDALSYMT IQQKEDITHS AQHALRLSAF GQIYKVLEMD PLPSSKPFQK YSWSVTDKEG AGSSALKRPF EDGLGDDKDP NKKMKRNLRK ILDSKAIDLM NALMRLNQIR PGLQYKLLSQ SGPVHAPVF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Involved in spermatogenesis and sperm function. Plays a role in regulation of cell growth. Binds to double-stranded DNA and RNA. Binds most efficiently to poly(I:C) RNA than to poly(dI:dC) DNA. Binds also to single-stranded poly(G) RNA. Binds non-specifically to the mRNA PRM1 3'-UTR and adenovirus VA RNA.

Specifications

Antigen STRBP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STRBP
Gene Accession No. Q96SI9
Gene Alias 6430510M02Rik; 74 kDa double-stranded RNA-binding protein; AI852513; C230082I21Rik; C86322; DKFZp434N214; double-stranded RNA-binding protein p74; epididymis luminal protein 162; FLJ11307; FLJ14223; FLJ14984; HEL162; ILF3L; MGC21529; MGC3405; p74; spermatid perinuclear RNA binding protein; spermatid perinuclear RNA-binding protein; SPNR; Strbp
Gene Symbols STRBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STRBP. Recombinant protein control fragment (Product #RP-102301).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less