missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRA8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580081
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Testis Tissue.
Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription.Chemical Identifiers
| STRA8 | |
| -20°C | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Rat | |
| STRA8 | |
| RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8 | |
| STRA8 | |
| Primary | |
| Antigen affinity chromatography |
| 500 μg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| 0 | |
| 500079 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of rat Stra8 (341-355aa DDDVMLLKCLETFDD). | |
| Antibody |
Specifications
| STRA8 | |
| Polyclonal | |
| Unconjugated | |
| STRA8 | |
| RGD1562852; stimulated by retinoic acid 8; stimulated by retinoic acid 8 homolog; stimulated by retinoic acid gene 8; stimulated by retinoic acid gene 8 homolog; stimulated by retinoic acid gene 8 protein; Stimulated by retinoic acid gene 8 protein homolog; stra8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 500079 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| 0 | |
| STRA8 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of rat Stra8 (341-355aa DDDVMLLKCLETFDD). | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |