Learn More
Thermo Scientific™ Storage Vials
Catalog No. B799515
Change view
Click to view available options
Color:
White
White, Clear
Closure Size:
13-425
15-425
18-400
20-400
Quantity:
100 Caps
100 Septa
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size Quantity
B799515 White, Clear 15-425 100 Septa
0339112E White 20-400 100 Caps
03394F White, Clear 13-425 100 Septa
03394B White, Clear 18-400 100 Septa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B799515 Supplier Thermo Scientific™ Supplier No. B799515
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Specifications
Description Septa for 15-425 Screw Cap
Color White, Clear
Material PTFE, Silicone
Quantity 100 Septa
Thickness (Metric) 2.6 mm
Closure Size 15-425
Product Type Septum Only
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.