Learn More
Accuform™ STOPOUT™ Ball Valve Lockout
Lockout devices control energy sources and ensure machine and equipment remain inoperable to prevent unexpected start-up or release of stored energy in accordance with OSHA 29 CFR 1910.147 requirements
Supplier:  Accuform™ KDD440
 Afficher plus de versions de ce produit
Numéro de catalogue. 19111149
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Works with ball valve handles either perpendicular or parallel to pipe and secures with padlock (sold separately)
  • Includes label printed with OSHA Danger header and message that reads LOCKED OUT with space to write more information as needed
  • Red octagon shape reinforces lockout status and conveys STOP
Spécifications
Spécifications
Ball Valve Lockout
Plastic
1/2 to 1/4 in.
1-1/2 in. to 2 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS