Learn More
Accuform™ STOPOUT™ Ball Valve Lockout
Lockout devices control energy sources and ensure machine and equipment remain inoperable to prevent unexpected start-up or release of stored energy in accordance with OSHA 29 CFR 1910.147 requirements
Supplier: Accuform™ KDD442
Description
- Works with ball valve handles either perpendicular or parallel to pipe and secures with padlock (sold separately)
- Includes label printed with OSHA Danger header and message that reads LOCKED OUT with space to write more information as needed
- Red octagon shape reinforces lockout status and conveys STOP
Specifications
| Ball Valve Lockout | |
| Plastic | |
| 5/8 to 1/4 in. | |
| 3/8 to 1-1/4 in. |