missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, Dried Powder, FCC, 86-89%, Spectrum™ Chemical

Catalog No. 18602710
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602710 50 kg Poly Bottle
18602707 500 g Poly Bottle
18602708 2.5 kg Poly Bottle
18602709 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602710 Supplier Spectrum Chemical Mfg Cor Supplier No. F106350KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F1063, 13463-43-9, FeSO4·xH2O

Spectrum™ Chemical Ferrous Sulfate, Dried Powder, FCC is used as an iron fortification food supplement additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13463-43-9
Molecular Formula FeH2O5S
Molecular Weight (g/mol) 169.92
InChI Key XBDUTCVQJHJTQZ-UHFFFAOYSA-L
IUPAC Name λ2-iron(2+) hydrate sulfate
SMILES O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline Powder or Needles
CAS Max % 100%
Quantity 50 kg
Packaging Poly Bottle
Percent Purity 86 to 89%
Grade FCC
Chemical Name or Material Ferrous Sulfate, Dried Powder