missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STIP1 Monoclonal Antibody (K92047_11E1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555875
Description
Sequence of this protein is as follows: AKRTYEEGLK HEANNPQLKE GLQNMEARLA ERKFMNPFNM PNLYQKLESD PRTRTLLSDP TYRELIEQLR NKPSDLGTKL QDPRIMTTLS VLLGVDLGSM DEEEEIATPP PPPPPKKETK PEPMEEDLPE NKKQALKEKE LGNDAYKKKD FDTALKHYDK AKELDPTNMT YITNQAAVYF EKGDYNKCRE LCEKAIEVGR ENREDYRQIA KAYARIGNSY FKEEKYKDAI HFYNKSLAEH RTPDVLKKCQ QAEKILKEQE RLAYINPDLA LEEKNKGNEC FQKGDYPQAM KHYTEAIKRN PKDAKLYSNR
Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).Chemical Identifiers
| STIP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521 | |
| Recombinant human STIP1 (immunogen range is 91-400). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| STIP1 |
| K92047_11E1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P31948 | |
| 10963 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| STIP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P31948 | |
| STIP1 | |
| Recombinant human STIP1 (immunogen range is 91-400). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K92047_11E1 | |
| Unconjugated | |
| STIP1 | |
| epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10963 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |