missing translation for 'onlineSavingsMsg'
Learn More

Stilbazo [Spectrophotometric reagent for Al and other metals], TCI America™

Catalog No. p-7118256
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50111G 1 g
A50115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50111G Supplier TCI America Supplier No. A50111G

Chemical Identifiers

CAS 1571-36-4
Molecular Formula C26H26N6O10S2
Molecular Weight (g/mol) 646.646
MDL Number MFCD00045752
InChI Key WMIFGUMCCUZRTH-SEPHDYHBSA-N
Synonym Stilbene-4,4′C-bis[(1-azo)-3,4-dihydroxybenzene]-2,2′C-disulfonic Acid Diammonium Salt
PubChem CID 87194021
IUPAC Name azane;5-[2-(3,4-dioxocyclohexa-1,5-dien-1-yl)hydrazinyl]-2-[(E)-2-[4-[2-(3,4-dioxocyclohexa-1,5-dien-1-yl)hydrazinyl]-2-sulfophenyl]ethenyl]benzenesulfonic acid
SMILES C1=CC(=C(C=C1NNC2=CC(=O)C(=O)C=C2)S(=O)(=O)O)C=CC3=C(C=C(C=C3)NNC4=CC(=O)C(=O)C=C4)S(=O)(=O)O.N.N

Specifications

Quantity 1 g
Formula Weight 646.65
Physical Form Crystalline Powder
Chemical Name or Material Stilbazo [Spectrophotometric reagent for Al and other metals]