accessibility menu, dialog, popup

UserName

CAS RN 1571-36-4

HNNHOOSOOOHHNNHOOSOOHONH3NH3(E/Z)

CAS RN 1571-36-4

IUPAC Name: 5-[2-(3,4-dioxocyclohexa-1,5-dien-1-yl)hydrazin-1-yl]-2-(2-{4-[2-(3,4-dioxocyclohexa-1,5-dien-1-yl)hydrazin-1-yl]-2-sulfophenyl}ethenyl)benzene-1-sulfonic acid diamine
Synonyms: 5-[2-(3,4-dioxocyclohexa-1,5-dien-1-yl)hydrazin-1-yl]-2-(2-{4-[2-(3,4-dioxocyclohexa-1,5-dien-1-yl)hydrazin-1-yl]-2-sulfophenyl}ethenyl)benzenesulfonic acid diamine
Molecular Weight (g/mol): 646.65
Molecular Formula: C26H26N6O10S2
InChi Key: WMIFGUMCCUZRTH-UHFFFAOYSA-N
SMILES: N.N.OS(=O)(=O)C1=C(C=CC2=C(C=C(NNC3=CC(=O)C(=O)C=C3)C=C2)S(O)(=O)=O)C=CC(NNC2=CC(=O)C(=O)C=C2)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results