missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stevia Leaves Extract, Sweet, Powder, 90%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S19645KG
Specifications
| 91722-21-3 | |
| 0.06 | |
| C44H70O23 | |
| HELXLJCILKEWJH-UHFFFAOYSA-N | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹â°.0â´,â¹]hexadecane-5-carboxylate | |
| 0.9 |
| 1 | |
| Poly Pail | |
| 5 kg | |
| CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O | |
| 967.02 | |
| Reagent |
Chemical Identifiers
| 91722-21-3 | |
| 967.02 | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹â°.0â´,â¹]hexadecane-5-carboxylate |
| C44H70O23 | |
| HELXLJCILKEWJH-UHFFFAOYSA-N | |
| CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O |
CAUTION: Read and understand the label and Safety Data Sheet (SDS) prior to use.