missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stevia Leaves Extract, Sweet, Powder, 90%, Spectrum™ Chemical
$254.54 - $27026.39
Chemical Identifiers
| CAS | 91722-21-3 |
|---|---|
| Molecular Formula | C44H70O23 |
| Molecular Weight (g/mol) | 967.02 |
| InChI Key | HELXLJCILKEWJH-UHFFFAOYSA-N |
| IUPAC Name | 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹â°.0â´,â¹]hexadecane-5-carboxylate |
| SMILES | CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18614086
|
Spectrum Chemical Mfg Cor
S196425GM |
25 g | Poly Bottle |
Each for $254.54
|
|
||||
|
18614087
|
Spectrum Chemical Mfg Cor
S1964100GM |
100 g | Poly Bottle |
Each for $484.89
|
|
||||
|
18614088
|
Spectrum Chemical Mfg Cor
S1964500GM |
500 g | Poly Pail |
Each for $1,467.61
|
|
||||
|
18614089
|
Spectrum Chemical Mfg Cor
S19645KG |
5 kg | Poly Pail |
Each for $5,720.11
|
|
||||
|
18614090
|
Spectrum Chemical Mfg Cor
S196425KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 91722-21-3 | |
| 967.02 | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹â°.0â´,â¹]hexadecane-5-carboxylate |
| C44H70O23 | |
| HELXLJCILKEWJH-UHFFFAOYSA-N | |
| CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O |
Specifications
| 91722-21-3 | |
| 0.06 | |
| C44H70O23 | |
| HELXLJCILKEWJH-UHFFFAOYSA-N | |
| 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹â°.0â´,â¹]hexadecane-5-carboxylate | |
| 0.9 |
| 1 | |
| Poly Bottle | |
| 25 g | |
| CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O | |
| 967.02 | |
| Reagent |
CAUTION: Read and understand the label and Safety Data Sheet (SDS) prior to use.