missing translation for 'onlineSavingsMsg'
Learn More

Stevia Leaves Extract, Sweet, Powder, 90%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1964500GM

 View more versions of this product

Catalog No. 18614088


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

91722-21-3
0.06
C44H70O23
HELXLJCILKEWJH-UHFFFAOYSA-N
3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹⁰.0⁴,⁹]hexadecane-5-carboxylate
0.9
1
Poly Pail
500 g
CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O
967.02
Reagent

Chemical Identifiers

91722-21-3
967.02
3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl 13-{[5-hydroxy-6-(hydroxymethyl)-3,4-bis({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxan-2-yl]oxy}-5,9-dimethyl-14-methylidenetetracyclo[11.2.1.0¹,¹⁰.0⁴,⁹]hexadecane-5-carboxylate
C44H70O23
HELXLJCILKEWJH-UHFFFAOYSA-N
CC12CCCC(C)(C1CCC13CC(=C)C(C1)(CCC23)OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1OC1OC(CO)C(O)C(O)C1O)C(=O)OC1OC(CO)C(O)C(O)C1O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: Read and understand the label and Safety Data Sheet (SDS) prior to use.