missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Steritop™ E-GP Funnel-less Sterile Vacuum Filtration Systems
GREENER_CHOICE

Catalog No. SEGPT0045
Change view
Click to view available options
Outlet Fitting:
38 mm threaded
45 mm threaded
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Outlet Fitting
SEGPT0045 45 mm threaded
SEGPT0038 38 mm threaded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SEGPT0045 Supplier MilliporeSigma™ Supplier No. SEGPT0045

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.

Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.

The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers.

SEGPT0038 is a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane that can directly connect to any volume, 45 mm receiver flask (glass or plastic) for sterilizing tissue culture media and buffers.

Specifications

Color Clear with blue adapter
Disposable Single-use
Material Polystyrene
Material (Membrane) PES Express PLUS
Pore Size 0.22 μm
Sterility Sterile
Filtration Area 40 cm²
For Use With (Application) Sterile filtration
Outlet Fitting 45 mm threaded
Outlet Style 45 mm threaded
Type Bottle Top Filter
Volume (Metric) <2 mL (limited by vial volume)
Diameter (Metric) 73 mm
Diameter (Metric) Filter 73 mm
Show More Show Less