missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Sterileware™ Upright Handle Ladles
Available in blue or white to allow for effective sample contrast, these sterile ladles are useful for sampling liquids and creams.
Supplier: Bel-Art™ F367750050
Description
- Upright handle allows for sampling from above container
- Curved handle end provides secure withdrawal of sample
- Fits into 2 in. openings such as a 45-gallon drum bunghole
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- White high density polyethylene (HDPE), or blue polypropylene (PP) ladles heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Chemical Identifiers
| Not Autoclavable | |
| 50 mL | |
| Polypropylene |
| 40/Pk. | |
| Ladle | |
| 26 cm |
Specifications
| 40/Pk. | |
| 50 mL | |
| Ladle | |
| 26 cm |
| Not Autoclavable | |
| Blue | |
| Polypropylene |