missing translation for 'onlineSavingsMsg'
Learn More

Water, USP Purified/nSTERILE Filtered, WFI Quality, Ricca Chemical

Catalog No. R919200055S
Change view
Click to view available options
Quantity:
20 L
200 L
Packaging:
Fiberboard Cube
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R919200055S 200 L Poly Drum
R919200020S 20 L Fiberboard Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R919200055S Supplier Ricca Chemical Company Supplier No. R919200055S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH ∼7
Color Clear, Colorless
Melting Point 0°C
CAS Min % 100
Boiling Point 100°C
CAS Max % 100
Conductivity ≤1.3 μS/cm at 25°C
Description WFI Quality
Endotoxin ≤0.25 EU/mL
UN Number 2853.90.9090
Percent Purity 100%
Grade USP
Packaging Poly Drum
Sterility Sterile
Density 1.00 g/cm3
Certificate of Analysis Yes
Quantity 200 L
Specific Gravity 1
Physical Form Liquid
Assay No assay needed
Chemical Name or Material Water
Show More Show Less
ShelfLife 12 Months
Recommended Storage Ambient/Room Temperature