missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Sterile Square HDPE Reagent Bottles with Wide Mouth Polypropylene Caps
Description
- Sterile reagent bottles - Sterile square HDPE bottles with polypropylene caps, designed for secure and versatile use in labs and industrial settings.
- Leak-proof design - Rigorously tested to prevent leaks under pressure, ensuring safe storage and transport of liquids and reagents.
- Excellent chemical resistance - Crafted from HDPE, offering high durability and compatibility with a variety of chemicals and reagents.
- Versatile lab and industrial use - Suitable for chemical storage, liquid handling, and mixing applications in research, laboratory, and industrial environments.
- Translucent and portable - Lightweight, translucent body for easy content visibility and a square design for optimal storage efficiency.
- Designed for chemical storage and transport, these leak-proof, chemical-resistant bottles are ideal for laboratory and industrial applications, -100°C to 120°C.
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Translucent |
| Material | HDPE |
| Sterility | Sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Product Type | Reagent Bottle |