Learn More
Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase pipetting performance with these sterile pipette tips, which provide accurate and precise performance due to superior tip straightness and a low coefficient of variation.
$645.97 - $963.20
Specifications
| Sterility | Sterile |
|---|---|
| Format | Rack |
| For Use With (Equipment) | Beckman Coulter Liquid Handling Systems |
| Catalog Number | Mfr. No. | Volume (Metric) | Packaging | Fit for Purpose | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Packaging | Fit for Purpose | Price | Quantity | |||
|
21-402-522
|
Thermo Scientific™
918011 |
20 μL | 50 x 96 tips | Sterile and PCR Ready |
Pack of 960 Each for $963.20
Case of 5 Pack for $4,816.00
|
|
|||
|
21-402-520
|
Thermo Scientific™
918251 |
20 μL | 10 x 96 tips | Sterile |
Pack of 960 Each for $645.97
Case of 5 Pack for $3,229.85
|
|
|||
|
21-402-525
|
Thermo Scientific™
919011 |
130 μL | 10 x 96 tips | Sterile and PCR Ready |
Pack of 960 Each for $943.29
Case of 5 Pack for $4,716.45
|
|
|||
|
21-402-523
|
Thermo Scientific™
919251 |
250 μL | 50 x 96 tips | Sterile |
Pack of 960 Each for $672.61
Case of 5 Pack for $3,363.05
|
|
|||
Description
Increase pipetting performance with Thermo ScientificTM Sterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:- Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
- Lot-tested during production on actual workstation for which they are created
- Extended length tips compatible with 96-, 384-, and 1536-DeepWell™ pates
- Low percentage coefficient of variation (%CV)
Specifications
| Sterile | |
| Rack | |
| Beckman Coulter Liquid Handling Systems |