missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems

Catalog No. 21402522
Change view
Click to view available options
Volume (Metric):
20 μL
130 μL
250 μL
Packaging:
10 x 96 tips
50 x 96 tips
Fit for Purpose:
Sterile
Sterile and PCR Ready
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Packaging Fit for Purpose
21402522 20 μL 50 x 96 tips Sterile and PCR Ready
21402520 20 μL 10 x 96 tips Sterile
21402525 130 μL 10 x 96 tips Sterile and PCR Ready
21402523 250 μL 50 x 96 tips Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21402522 Supplier Thermo Scientific™ Supplier No. 918011

Increase pipetting performance with these sterile pipette tips, which provide accurate and precise performance due to superior tip straightness and a low coefficient of variation.

Increase pipetting performance with Thermo ScientificTM Sterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Color Black
Format Rack
Tip Style Conductive
Packaging 50 x 96 tips
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile and PCR Ready