missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec Sterile CyQuanol for Canada
The CyQuanol™ is a ready-to-use blend of quaternary ammonium and ethyl alcohol. Health Canada registered with a 1-minute contact time, it’s effective against bacteria, viruses, fungi, and TB.
$625.48 - $726.34
Specifications
| Product Type | Cleaner and Disinfectant Solution |
|---|---|
| Form | Liquid |
| Scent | Original |
| Sterility | Sterile |
| Composition | Quaternary Ammonium/Ethyl Alcohol |
Description
- 0.2 micron filtered and appropriate for use in critical environments
- Low residues that are easily removed with common solvents
- 1-minute contact time to easily maintain compliance and appropriate dwell time
- Double bagged in linear tear packaging making it easy to open even when wearing gloves
- Available gamma irradiated to a Sterility
- Assurance Level of 10-6 SAL making it suitable for Grade A/B (ISO 5) environments
- Trigger sprayer or reclosable cap for better control of solvent usage
- Bleach-free and compatible with common cleanroom surfaces
Specifications
| Cleaner and Disinfectant Solution | |
| Original | |
| Quaternary Ammonium/Ethyl Alcohol |
| Liquid | |
| Sterile |