missing translation for 'onlineSavingsMsg'
Learn More

Contec Sterile CyQuanol for Canada

Catalog No. 17103020
Change view
Click to view available options
Appearance:
Colorless
Colorless to Slightly Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Appearance
17103020 Colorless
17103019 Colorless to Slightly Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17103020 Supplier Contec Supplier No. SBT09CQCA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The CyQuanol™ is a ready-to-use blend of quaternary ammonium and ethyl alcohol. Health Canada registered with a 1-minute contact time, it’s effective against bacteria, viruses, fungi, and TB.

  • 0.2 micron filtered and appropriate for use in critical environments
  • Low residues that are easily removed with common solvents
  • 1-minute contact time to easily maintain compliance and appropriate dwell time
  • Double bagged in linear tear packaging making it easy to open even when wearing gloves
  • Available gamma irradiated to a Sterility
  • Assurance Level of 10-6 SAL making it suitable for Grade A/B (ISO 5) environments
  • Trigger sprayer or reclosable cap for better control of solvent usage
  • Bleach-free and compatible with common cleanroom surfaces

Specifications

Product Type Cleaner and Disinfectant Solution
Form Liquid
Scent Original
Sterility Sterile
Appearance Colorless
Composition Quaternary Ammonium/Ethyl Alcohol