missing translation for 'onlineSavingsMsg'
Learn More

Sterigmatocystin Solution, 50 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018206
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018206 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018206 Supplier Merck Emd Millipore Supplier No. 329861ML

Sterigmatocystin is classified under the mycotoxin family of compounds. It is usually produced by the fungi of several species of Aspergillus.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 10048-13-2
Flash Point 6°C - Closed Cup
Linear Formula C18H12O6
MDL Number MFCD00005084
Synonym 3 A,12 c-Dihydro-8-hydroxy-6-methoxy-7 H-furol[3 ′,2 ′:4,5 ]furo[2,3-c]xanthen-7-one
Type Analytical Standard
Quantity 1 mL
Formula Weight 324.28
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in acetonitrile