missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Stericup™ Quick Release Bottle Top Receiver Flasks

Catalog No. S200B05RE
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S200B05RE 500 mL
S200B02RE 250 mL
S200B10RE 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S200B05RE Supplier MilliporeSigma™ Supplier No. S200B05RE
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.

Steritop™ bottle top filter units provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33 mm) or wide-mouth (45 mm) bottles.

A small tab inside the funnel secures an optional prefilter in-place during filtration.

For added protection against contamination, the threads for bottle attachment have been slightly recessed in case the funnel accidentally touches a work surface.

Specifications

Outlet Fitting 45 mm
Volume (Metric) 500 mL