missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Stericup™ Quick Release-GP Sterile Vacuum Bottle Top Filtration Systems

Catalog No. S2GPU11RE
Change view
Click to view available options
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
Volume (Metric) Receiver:
150 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Volume (Metric) Receiver
S2GPU11RE 1000 mL 1000 mL
S2GPU01RE 150 mL 150 mL
S2GPU02RE 250 mL 250 mL
S2GPU05RE 500 mL 500 mL
S2GPU10RE 1000 mL 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S2GPU11RE Supplier MilliporeSigma™ Supplier No. S2GPU11RE
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device.

The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop™ bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Stable base and easy grip design of the Stericup™ device enable any user to comfortably perform all stages of the workflow in a laminar flow hood.
  • Recessed bottom of Stericup™ receiver flasks allows flasks to be stacked easily in refrigerated spaces.

Applications

  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.

Specifications

Color Clear with blue adapter
Disposable Single-use
Filter Type Membrane Filter
Material PES
Material (Membrane) Polystyrene
Pore Size 0.22 μm
Sterility Sterile
For Use With (Application) Sterile filtration
Type Bottle Top Filter
Volume (Metric) 1000 mL
Volume (Metric) Receiver 1000 mL
Length (Metric) 319 mm
Height (Metric) 319 mm
Diameter (Metric) 13.5 cm
Diameter (Metric) Filter 73 mm
Show More Show Less