Learn More
MilliporeSigma™ Stericup™ E-GP Funnel-less Sterile Vacuum Filtration Systems
GREENER_CHOICE
The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.
Supplier:  MilliporeSigma™ SEGPU0545
 View more versions of this product
Catalog No. SEGPU0545
Get 1 Extra Pack of Filter Units When You Buy 3
View all promotions
Only null left
Description
Description
Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers. Like Stericup Quick Release filters, Stericup E devices feature a tactile-stop cap to signal secure closure, an ample textured writing surface, and filter membrane characteristics clearly labeled on the filter collar.SEGPU0538 combines a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane, with a 500 mL receiver flask for sterilizing tissue culture media and buffers. Not recommended for reagents prepared in other vessel type devices like flasks or anything without a threaded neck.
Specifications
Chemical Identifiers
Clear with blue adapter
Polystyrene
0.22 μm
40 cm²
Bottle Top Filter
73 mm
Single-use
PES Express PLUS
Sterile
Sterile filtration
73 mm
Specifications
Clear with blue adapter
Polystyrene
0.22 μm
40 cm²
45 mm threaded
Bottle Top Filter
73 mm
Single-use
PES Express PLUS
Sterile
Sterile filtration
45 mm threaded
500 mL
73 mm
Product Suggestions
Product Suggestions