Learn More
Stoelting™ Stereotaxic Robot Drill and Microinjector Robot
Stoelting Stereotaxic Robot Drill and Microinjector Robot provides for a fully software driven solution to stereotaxic surgery
$104,705.70
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 1
Catalog Number Mfr. No. Animal Price Quantity & Availability  
Catalog Number Mfr. No. Animal Price Quantity & Availability  
10000190
View Documents Promotion Details
Stoelting™
51705
Mouse N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Allows for high precision injection.
  • After loading the syringe, the injection robot allows for multiple injections in different animals without tool exchange or recalibration
  • Completely automated solution for stereotaxic surgery
  • Features:
    • High Throughput Drill and Inject
    • No Tool Exchange
    • Define/Store Protocols
    • Precise Depth Control
    • Brain Windowing
    • Contour Drilling
    • Smooth Edges
    • Skull Thinning
Specifications
Specifications
Neuroscience