Learn More
Stoelting™ Stereotaxic Robot Drill and Microinjector Robot
Description
- Allows for high precision injection.
- After loading the syringe, the injection robot allows for multiple injections in different animals without tool exchange or recalibration
- Completely automated solution for stereotaxic surgery
- Features:
- High Throughput Drill and Inject
- No Tool Exchange
- Define/Store Protocols
- Precise Depth Control
- Brain Windowing
- Contour Drilling
- Smooth Edges
- Skull Thinning
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Animal | Mouse |