missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ StepOnePlus™ Real-Time PCR System Upgrade
Description
Includes
Upgrade
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Thermal Accuracy | 0.25°C (35°C to 95°C) of Display Temperature |
| Sensitivity | 1 Copy |
| Dynamic Range | Linear Dynamic Range greater than 9 Log Units (Detection) |
| Content And Storage | Upgrade |
| Thermal Uniformity | 0.25°C (35°C to 95°C) of Setpoint/Display Temperature |
| Height (Metric) | 51.2 cm |
| Depth (Metric) | 42.7 cm |
| Thermal Range | 4°C to 100°C |
| For Use With (Application) | Genotyping and Genomic Profiling, Real Time PCR (qPCR), Real Time PCR-based Gene Expression Profiling |
| Description | Scan Time: <2 hr./run (Standard Mode), <40 min./run (Fast Mode) |
| Show More |