Learn More
Applied Biosystems™ StepOne™ Real-Time PCR System, laptop
Description
Includes
Laptop Computer
The StepOne™ Real-Time PCR System can be setup in a variety of configurations and comes ready to use, out of the box, with intuitive data analysis and instrument control software. Utilizing robust LED based 3-color optical recording, the StepOne™ Real-Time PCR System is designed to deliver precise, quantitative Real-Time PCR results for a variety of genomic research applications. Includes a laptop computer.
Features of the The StepOne™ Real-Time PCR System include:
- Advanced software and instrumentation for performing a wide array of genomic assays.
- Sensitive 3-color optical LED recording syste
- Intuitive and robust software perfect for both first-time and advanced users
- Simple and flexible instrument set-up and usage
StepOne™ Software:
The StepOne™ Software included with the StepOne™ Real-Time PCR System runs on Windows™ XP, Window Vista™, and Windows™ 7 operating systems and provides instrument control, data collection, and data analysis capabilities. This latest version includes capabilities to collect melt curve data for High Resolution Melt (HRM) applications and the option to export in Real-Time PCR Data Markup Language (RDML) for compatibility with MIQE guidelines.
Rapid and Simple Assay Set-Up:
This remarkably simple Real-Time PCR system is designed with an LCD touchscreen and a user-friendly interface that brings the power of genetic studies to researchers new to Real-Time PCR. The intuitive software and protocol wizards help guide new users through their Real-Time PCR experiments. The StepOne™ Real-Time PCR System also includes a quick-start setup so that you start a run immediately and enter plate information at a later time.
Sensitive LED-Based 3-Color Fluorescence Reading:
The StepOne™ Real-Time PCR System utilizes a long-life LED-based optical system that can record fluorescence from FAM™/SYBR™ Green, VIC™/JOE™, and ROX™ dyes. This cost-effective, 3-color, 48-well system delivers precise, quantitative Real-Time PCR results and saves data from all filters in every run without depending on a computer or plate setup. It can discriminate between 2 populations of 5,000 and 10,000 template copies of a TaqMan™ assay with 99.7% confidence.
Compatible with Many Genomic Analysis Techniques:
Perform a variety of standard and demanding genetic analysis research techniques on one instrument using the user friendly StepOne™ Real-Time PCR System and software. The StepOne™ System supports many Real-Time PCR applications, including the following:
- SNP Genotyping
- Gene Expression Analysis
- MicroRNA Expression
- Translocation Analysis
- Gene Detection
- Viral Load Analysis
- Standard Curve (absolute quantitation)
- Relative Standard Curve
- Comparative Ct (ΔΔCt) (relative quantitation)
- Genotyping and Presence/Absence
- Melt Curve Analysis
- High Resolution Melting (additional software required)
Versatile Instrument Configuration Options:
The ultra-compact footprint of a StepOne™ System can be installed in multiple distinct configurations, providing unmatched flexibility and convenience that can allow a fit in any laboratory. The StepOne™ Real-Time PCR System can be used via a PC, with a PC connected to a Local Area Network (LAN), or as a stand-alone instrument (PC-free). Each StepOne™ Real-Time PCR System is factory-calibrated for optical and thermal accuracy, so simply remarkable Real-Time PCR results can be obtained right out of the box.
Real-Time Data Monitoring, Dissemination, and Storage:
The system measures amplification as it occurs, allowing you to monitor the progress of your experiment cycle by cycle, either on the machine or remotely. Your data is stored on the instrument itself, and can be viewed and stored automatically via remote access or transferred via email or a USB flash drive. Data can also be conveniently exported as PowerPoint™, Excel™, and graphical image files.
Expanded Gene Expression Capabilities:
The advanced software provided with the StepOne™ Real-Time PCR System now includes the powerful Gene Expression Study Package. This software package allows for greater flexibility and accuracy in your gene expression assays through:
- Analysis of an unlimited number of plates in one study
- Sorting of data by biological or technical replicate group
- Use of multiple endogenous controls
- Assay efficiency correction
High Resolution Melting Capability:
Now with High Resolution Melt Software v3.0 you can perform post-PCR sequence variation analysis with the StepOne™ Real-Time PCR System. This separately purchased software simplifies set-up by maintaining assay specific settings and accepts pasted plate layout information directly from Excel. HRM Software v3.0 also has an improved clustering algorithm for increased sensitivity and accuracy, and the ability to conduct separate analyses of multiple assays run on a single plate.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
Specifications
| Format | 48-well Plate, 0.1 mL Tubes |
| Thermal Accuracy | ± 0.25°C (35°C to 95°C) of Display Temperature |
| Sample Ramp Rate | Fast Mode: ± 2.2°C/s, Standard mode: ± 1.6°C/s |
| Sensitivity | 1 Copy |
| Temperature Range (Metric) | 4 to 100°C |
| Reaction Volume Range | 10–30 μL (Standard Curve Experiments: 40 μL in Standard Mode is Validated) |
| Passive Reference Dye | ROX (Separate Tube), No ROX, ROX (Pre-mixed) |
| Dynamic Range | Linear Dynamic Range greater than 9 Log Units (Detection) |
| Content And Storage | Laptop Computer |
| Calibrated Dye | VIC™, SYBR™ Green I, JOE™, FAM™, ROX™ |
| Show More |
Frequently Asked Questions (FAQs)
This issue can occur if the run is interrupted. To prevent this from happening in the future, avoid ending the PCR with an infinite hold stage or interrupting the run. Please send the file to techsupport@thermofisher.com so that our software group can attempt to recover the data.
This is generally caused by incorrect placement of tube strips, resulting in a sample block that can't properly be lowered. We recommend contacting Instrument Services to have an engineer walk you through a manual drawer-lowering process during instrument power-up. To prevent this from happening in the future, place tubes toward the middle of the block or use a placeholder strip on the opposing side to counterbalance block pressure.
This issue can occur if the Start Run button is clicked more than once or if illegal characters are used in the experiment/file name. You can recover the data yourself by overriding the calibration file with one that represents the calibration status of the instrument when you ran that particular file. Use the toolbar at the top to navigate: Analysis then Override Calibration then Use Calibration From Another File.
This issue can occur if duplicate samples are added to the experiment, if illegal characters are used in the experiment/file name, or if the file extension was changed to .eds from a different extension.
Please send the file to techsupport@thermofisher.com to attempt recovery.
This error means that the data collection was not turned on as appropriate. If this is inadvertently turned off, no data will be collected. Thus, the data are not retrievable in such a file.