Learn More
Gibco™ StemPro™ hESC SFM
StemPro hESC SFM is a serum- and feeder-free medium specially formulated to support the growth and expansion of pluripotent stem cells (hESCs and iPSCs). It has been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities while maintaining differentiation potential.
Supplier: Gibco™ A1000701
Description
StemPro hESC SFM is a serum- and feeder-free medium specially formulated to support the growth and expansion of pluripotent stem cells (hESCs and iPSCs). It has been shown to support PSC growth for >50 passages without any signs of karyotypic abnormalities while maintaining differentiation potential.
StemPro hESC SFM has been extensively tested and is proven to maintain pluripotency in a growing list of hESC lines, including BG01, BG02, BG03, H1, H9, and BG01V. In addition, it has been used to scale up production of hESCs to over 1 x 1010 cells. StemPro hESC SFM has been shown to support hESC and iPSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of hESCs to differentiate into all three germ line lineages.
StemPro hESC SFM components
- StemPro hESC Supplement
- DMEM/F-12 with GlutaMAX medium
- Bovine serum albumin 25% (BSA)
Additional components required for complete medium (need to be purchased separately)
- FGF BASIC (full length) REC HU (Cat. no. PHG0261)
- 2-Mercaptoethanol (Cat. no. 21985-023)
Applications
Formulated for the growth and expansion of human embryonic and human induced pluripotent stem cells.
Specifications
| Embryonic Stem Cells (ESCs) | |
| Liquid | |
| Serum-free | |
| cGMP for medical devices, 21 CFR Part 820 and ISO 13485 | |
| 1 Kit | |
| Human | |
| Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic) |
| Serum-free | |
| Stem Cell Media | |
| GlutaMAX, Phenol Red | |
| StemPro | |
| Supplement - dry ice, Basal - ambient, BSA - ambient | |
| • StemPro™ hESC Supplement, store at -5 to -20°C and protect from light • DMEM/F-12 with GlutaMAX medium, store at 2 to 8°C and protect from light • Bovine serum albumin (BSA) 25%, store at 2 to 8°C and protect from light |
For Research Use Only. Not for use in diagnostic procedures.