missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ Adipogenesis Differentiation Kit

Catalog No. A1007001
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1007001 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1007001 Supplier Gibco™ Supplier No. A1007001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

StemPro™ Adipogenesis Differentiation Kit

For Complete Differentiation of Human Mesenchymal Stem Cells into Fat Cells

StemPro™ Adipogenesis Differentiation Kit has been developed for the adipogenic differentiation of mesenchymal stem cells (MSCs) in tissue culture vessels. The kit contains all reagents required for inducing MSCs to be committed to the adipogenesis pathway and generate adipocytes. Using StemPro™ Adipogenesis Differentiation Kit in combination with StemPro™ MSC SFM or MesenPRO RS™ Medium provides a standardized culture workflow solution for MSC isolation, expansion and differentiation into lipid vesicle-forming adipocytes.

Specifications

Cell Type Stem Cells (Mesenchymal)
Content And Storage This kit is shipped in two parts with separate storage requirements. These components are not sold individually:
• StemPro™ Adipocyte Differentiation Basal Medium (100 ml; store in the dark at 2°C to 8°C)
• StemPro™ Adipogenesis Supplement (10 ml; store in the dark at -5°C to -20°C)

The Basal Medium ships at room temperature, while the Supplement ships on dry ice.
Culture Environment CO2
For Use With (Application) Stem Cell Differentiation
Product Type Adipogenesis Differentiation Kit
Endotoxin Level ≤50 EU/mL, ≤ 50 EU/ml
Format Liquid
Glucose High Glucose
Glutamine GlutaMAX™-I
HEPES Buffer No HEPES
Inorganic Salts Calcium, Sodium Phosphate
Phenol Red Indicator No Phenol Red
Product Line Gibco, StemPro
Purity or Quality Grade Research Grade
Quantity 1 Kit
Serum Level Standard Serum
Sodium Bicarbonate Buffer Sodium Bicarbonate
Sodium Pyruvate Additive Sodium Pyruvate
Species Human
Supplements Adipogenesis Supplements
Show More Show Less
What kinds of growth factors are used in mesenchymal stem cell (MSC) research?

Growth factors used for MSC maintenance and self-renewal: LIF, FGF-basic, EGF, HGF, PDGF, and Wnt3.

Growth factors or cytokines for MSC differentiation: BMP-2and TGF-β1.

(Arthritis Res Ther 9:204 (2007)

View our Gibco™ MSC research products.

How can I culture mesenchymal stem cells (MSCs) in vitro?

MSCs may be cultured in either serum-containing medium or serum-free medium. The standard culturing conditions are DMEM, low glucose with 10% FBS. MSCs can also be grown in reduced-serum (2%) Gibco MesenPRO RS Medium (Cat. No. 12746-012), or in serum-free Gibco StemPro MSC SFM XenoFree Medium (Cat. No. A10675) and Gibco StemPro MSC SFM (Cat. No. A10332). In general, MSCs grow better under hypoxic conditions (2% O2).

How do you characterize human mesenchymal stem cells (MSCs)?

Human MSCs can be identified by flow cytometry, typically displaying CD73+, CD90+, CD105+, CD11b-, and CD45- marker characteristics (Blood 109:4245 (2007)). Human MSC expresses Oct4, Sox2, and Rex-1; these may be verified using RT-PCR (Arthritis Res Ther 9:204 (2007)).

What are mesenchymal stem cells?

Mesenchymal stem cells (MSCs) are multipotent cells isolated primarily from bone marrow or fat tissues that exhibit the ability to differentiate into bone, cartilage, and fat cells. Under normal cell culture conditions, MSCs isolated from bone marrow are spindle shaped with the unique ability to adhere to uncoated plastic culture dishes (Arthritis Res Ther 9:204 (2007)).

For Research Use Only. Not for use in diagnostic procedures.