missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Stem Cell Qualified ECM Gel
Stem cell qualified basement membrane extract (BME) for human ES and iPS cell culture.
Supplier: MilliporeSigma™ CC1315ML10P
Description
- Stem Cell Qualified ECM Gel is a soluble basement membrane extract (BME) purified from Engelbreth-Holm-Swarm (EHS) tumor that has been pre-qualified for human ES/iPS cell culture
- The matrix polymerizes at 20 to 40°C to form a reconstituted basement membrane
- ECM Gel contains laminin, collagen IV, entactin and heparan sulfate among other ECMs
- The matrix has been extensively tested and optimized for human ES/iPS cell culture
- Eliminates the need for time-consuming prescreening and lot identification
- Provides an optimized ECM coating necessary for long term culture of pluripotent human ES/iPS cells
- Supports feeder-free culture of human ES/iPS cells in serum-free media
- Compatible with all feeder free human ES/iPS cell expansion media (i.e. mTeSR™, PluriSTEM™)
Specifications
| Gel | |
| 10 x 5 mL | |
| < 20 EU/mL | |
| Store at -20°C, thaw overnight on ice in a 2 to 8°C refrigerator |
| Human ES and iPS cell culture | |
| < 5% spontaneous differentiation for 3 passages using Human iPS cells | |
| Red, clear of particulates | |
| Basement membrane extract (BME) |