Learn More
Diversified Spaces™ Steel Workbench DFS Item
Constructed from heavy gauge steel members that can be combined in several ways.
$4654.40 - $4654.40
Specifications
Frame Material Heavy-gauge Steel
Shelf Type 3/4-deep Lower Shelf
Top Style 1.75 in. maple
Product Type Workbench
View More Specs
Products 1
Catalog Number Mfr. No. Dimensions (D x W x H) Top Style Price Quantity  
Catalog Number Mfr. No. Dimensions (D x W x H) Top Style Price Quantity  
36-104-4453 DFS Item
View Documents
Diversified Spaces™
MLB1116
30 x 96 x 32 in. 1.75 in. maple
Each for $4,654.40
Only null left
Description
Description
  • Constructed of heavy-gauge steel frame
  • Welded-construction legs with floor feet
  • Three-quarter-deep lower shelf
  • Rear stringer for added strength
  • 12-gauge steel top wrapped over 2 in. thick core
Specifications
Specifications
Heavy-gauge Steel
3/4-deep Lower Shelf
1.75 in. maple
Workbench
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'