missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid 98.0+%, TCI America™

Catalog No. p-7135837
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S016325G 25 g
S0163500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S016325G Supplier TCI America Supplier No. S016325G
Only null left

Chemical Identifiers

CAS 57-11-4
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
Synonym stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
PubChem CID 5281
ChEBI CHEBI:28842
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 70°C
Color White
Boiling Point 240°C
Quantity 25 g
Formula Weight 284.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Stearic Acid
EINECSNumber (2)-0608
RTECSNumber WI2800000
TSCA Yes