missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearic Acid, Free Acid, 99%, MP Biomedicals™
$684.63
Chemical Identifiers
| CAS | 57-11-4 |
|---|---|
| Molecular Formula | C18H36O2 |
| Molecular Weight (g/mol) | 284.48 |
| MDL Number | MFCD00002752 |
| InChI Key | QIQXTHQIDYTFRH-UHFFFAOYSA-N |
| Synonym | stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid |
| PubChem CID | 5281 |
| ChEBI | CHEBI:28842 |
| IUPAC Name | octadecanoic acid |
| SMILES | CCCCCCCCCCCCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10111780
|
N/A | 100 g |
Each for $684.63
|
|
|||||
Chemical Identifiers
| C18H36O2 | |
| MFCD00002752 | |
| stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid | |
| CHEBI:28842 | |
| CCCCCCCCCCCCCCCCCC(O)=O |
Specifications
| 57-11-4 | |
| 99% | |
| MFCD00002752 | |
| QIQXTHQIDYTFRH-UHFFFAOYSA-N | |
| octadecanoic acid | |
| 5281 | |
| Powder |
| White to Yellow | |
| C18H36O2 | |
| stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid | |
| CCCCCCCCCCCCCCCCCC(O)=O | |
| 284.48 | |
| CHEBI:28842 | |
| Stearic Acid, Free Acid |