missing translation for 'onlineSavingsMsg'
Learn More

Stearanilide 98.0+%, TCI America™

Catalog No. S031425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031425G 25 g
S0314100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S031425G Supplier TCI America Supplier No. S031425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-54-7
Molecular Formula C24H41NO
Molecular Weight (g/mol) 359.60
MDL Number MFCD00048482
InChI Key ZOLJFBQEKSZVCB-UHFFFAOYSA-N
Synonym N-Phenylstearamide, N-Stearoylaniline
PubChem CID 69476
IUPAC Name N-phenyloctadecanamide
SMILES CCCCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1

Specifications

Melting Point 94°C
Color White
Boiling Point 154°C
Quantity 25 g
Formula Weight 359.60
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Stearanilide
EINECSNumber (3)-2698
TSCA Yes