missing translation for 'onlineSavingsMsg'
Learn More

Stearamide 90.0+%, TCI America™

Catalog No. S007525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S007525G 25 g
S0075250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S007525G Supplier TCI America Supplier No. S007525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-26-5
Molecular Formula C18H37NO
Molecular Weight (g/mol) 283.50
MDL Number MFCD00008038
InChI Key LYRFLYHAGKPMFH-UHFFFAOYSA-N
Synonym stearamide, stearylamide, octadecylamide, stearoylamide, stearoylamine, octadecamide, stearic acid amide, stearic amide, kemamide s, adogen 42
PubChem CID 31292
ChEBI CHEBI:34900
IUPAC Name octadecanamide
SMILES CCCCCCCCCCCCCCCCCC(N)=O

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 25 g
Formula Weight 283.50
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stearamide
EINECSNumber (2)-0824
RTECSNumber RG0182000
TSCA Yes