accessibility menu, dialog, popup

UserName

H3CNH2O

CAS RN 124-26-5

IUPAC Name: octadecanamide
Synonyms: stearamide
Molecular Weight (g/mol): 283.5
Molecular Formula: C18H37NO
InChi Key: LYRFLYHAGKPMFH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results