Learn More
StatLab StatPrep Preserve Prefill
Cytology preservative.
Supplier:  StatLab CY50NONGYN20ML
 View more versions of this product
Catalog No. CY50NONGY20
Only null left
Explore available promotions
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Description
Description
  • StatPrep™ Preserve is used for the preservation and transport of cytology specimens intended for liquid-based cytology processing
Specifications
Specifications
Preservative
Not Autoclavable
No
20 mL Non-gynecological Cytology Prefills
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Pre-filled container