Learn More
StatLab StatPrep Preserve Prefill
Cytology preservative.
$811.96
Specifications
| Type | Preservative |
|---|---|
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |
| Reusable | No |
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|
CY50GYN20ML
|
StatLab
CY50GYN/20ML |
20 mL Gynecological Cytology Prefills |
Case of 100 Each for $811.96
|
|
|||||
|
CY50NONGY20
|
StatLab
CY50NONGYN/20ML |
20 mL Non-gynecological Cytology Prefills |
Case of 100 Each for $811.96
|
|
|||||
Description
- StatPrep™ Preserve is used for the preservation and transport of cytology specimens intended for liquid-based cytology processing
Specifications
| Preservative | |
| Not Autoclavable | |
| Non-sterile | |
| No |
Pre-filled container